Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Imidazole-2-carboxylic acid ethyl ester

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Ethyl imidazole-2-carboxylateEthyl 1H-imidazole-2-carboxylate

OR3003-25G OR3003-5G OR3003-1G
76573-458EA 140 USD InStock
76573-458 76573-454 76573-456
Imidazole-2-carboxylic acid ethyl ester
Imidazole-2-carboxylic acid ethyl ester


Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Boiling Pt: 48…51 °C/0.3mmHg
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD03426031
CAS Number: 33543-78-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty