Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium-2-naphthalenesulfonate 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Sodium 2-naphthalenesulfonateSodium 2-naphthalenesulphonate2-Naphthalenesulphonic acid Sodium Salt2-Naphthalenesulfonic acid Sodium Salt

OR30530-250G OR30530-1KG OR30530-5KG
76574-412EA 183 USD InStock
76574-412 76574-410 76574-408
Sodium-2-naphthalenesulfonate 95%
Sodium-2-naphthalenesulfonate


Formula: C₁₀H₈O₃S
MW: 230.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064186
CAS Number: 532-02-5
EINECS: 208-523-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty