Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromothieno[2,3-c]pyridine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Bromo-6-aza-1-benzothiophene3-Bromo-6-azabenzo[b]thiophene

OR30581-1G OR30581-100MG
76574-536EA 129 USD InStock
76574-536 76574-534
3-Bromothieno[2,3-c]pyridine
3-Bromothieno[2,3-c]pyridine


Formula: C₇H₄BrNS
MW: 214.082 g/mol
Melting Pt: 97…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11052627
CAS Number: 28783-17-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty