Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,6-Dichloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

OR30659-250MG OR30659-1G
76574-614EA 172 USD InStock
76574-614 76574-616
4,6-Dichloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine
4,6-Dichloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine


Formula: C₇H₆Cl₂N₄
MW: 217.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11869749
CAS Number: 1072895-86-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty