Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Aminoethyl)-1-boc-azetidine 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate1-Boc-(3-Aminoethyl)azetidine1-Azetidinecarboxylic acid, 3-(1-aminoethyl)-, 1,1-dimethylethyl ester3-(1-Aminoéthyl)-1-azétidinecarboxylate de 2-méthyl-2-propanyle

OR308034-250MG OR308034-1G
76575-126EA 24 USD InStock
76575-126 76575-128
3-(2-Aminoethyl)-1-boc-azetidine 97%
3-(2-Aminoethyl)-1-boc-azetidine


Formula: C₁₀H₂₀N₂O₂
MW: 200.282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878606
CAS Number: 898271-20-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty