Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-Thz-OH 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Boc-L-thiazolidine-4-carboxylic acid

OR8031-100G OR8031-25G OR8031-5G
76563-230EA 301.79 USD InStock
76563-230 76559-546 76559-666
Boc-Thz-OH 98%
Boc-Thz-OH


Formula: C₉H₁₅NO₄S
MW: 233.282 g/mol
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00077002
CAS Number: 51077-16-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty