Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluoro-2-nitrobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC1745-1G PC1745-5G PC1745-25G
76561-084EA 89 USD InStock
76561-084 76560-686 76562-154
5-Fluoro-2-nitrobenzonitrile 98%
5-Fluoro-2-nitrobenzonitrile


Formula: C₇H₃FN₂O₂
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD01632196
CAS Number: 50594-78-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty