Would you like to log in again?
Eisco Rubber Dropper Bulbs
Supplier: EISCO SCIENTIFIC LLC
CH0232B
470345-416EA 0.49 USD InStock
470345-416
Eisco Rubber Dropper Bulbs
Pipettes Pipette Bulbs
For use with dropper pipettes.
  • Heavy-duty red rubber
  • Fits pipettes and medicine droppers measuring 5 to 6 mm in diameter
  • Measures 1.5H×0.75" Ø
Bulbs made of quality, heavy-duty red rubber with a 2 ml capacity. Fits pipettes and medicine droppers measuring 5 to 6 mm in diameter. Each bulb measures 1.5" tall, and 0.75" in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty