Would you like to log in again?
Eisco Natural Fiber Test Tube Brushes with Fan-Shaped Ends
Supplier: EISCO SCIENTIFIC LLC
CH0204APK12 CH0204BPK12 CH0204FPK12
470345-440EA 25.2 USD InStock
470345-440 470344-336 470343-982
Eisco Natural Fiber Test Tube Brushes with Fan-Shaped Ends
Brushes
Made for scrubbing test tubes, culture tubes, and pipes.
  • Designed to scrub narrow glassware
  • Fanned end that allows you to easily clean rounded surfaces and corners
  • Moderate flexibility allows easy modification to handle shape
These semi-stiff fiber bristle brushes are perfect for scrubbing narrow tube-shaped glassware such as test tubes and culture tubes. Each brush features a fanned end that allows you to easily clean rounded surfaces and corners. Each brush has a stainless steel, twisted handle that is semi-flexible, allowing you to modify the shape as needed. Each handle has a looped end, for easy hanging.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty