Would you like to log in again?
Pro-Wipe® SE Knit All-Purpose Sealed Edge Knit Wipes, Berkshire
Supplier: Berkshire
PWSEK09.14
76424-330CS 416.98 USD InStock
76424-330
Pro-Wipe® SE Knit All-Purpose Sealed Edge Knit Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Pro-Wipe® SE knit are industrial cleanroom wipes.
  • Chemically compatible with common cleaning and disinfecting solutions
  • Superior durability and strength
  • Ultrasonically sealed edge
  • Pre-laundered wipe reduces particle contamination
  • Durable and versatile wipe suited for controlled environments use
  • Autoclavable
Pro-Wipe® SE knit is an all-purpose sealed edge knit wipe. It is cleanroom laundered for use in controlled environment and industrial applications. This is a durable wipe with exceptional cleanliness compared to other industrial wipes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty