Would you like to log in again?
Eisco Nylon Cylinder Brushes with Fan-Shaped End
Supplier: EISCO SCIENTIFIC LLC
CH0220F CH0220E
470343-618EA 10.7 USD InStock
470343-618 470345-736
Eisco Nylon Cylinder Brushes with Fan-Shaped End
Brushes
Made for cleaning graduated cylinders, bottle necks, and more.
  • Medium-soft nylon bristles
  • Stainless steel, twisted wire handle that is resistant to corrosion
  • Fanned end for reaching every surface and corner
  • Moderate flexibility allows easy modification to handle shape
This medium soft nylon bristle brush is perfect for cleaning hard to reach, narrow spaces such as graduated cylinders, bottle necks, and more. The fanned end allows you to easily clean rounded surfaces and reach all corners. The stainless steel, twisted handle is semi-flexible, allowing you to modify the shape as needed. Looped end for easy hanging.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty