Would you like to log in again?
The Wizard® Genomic Plant DNA Purification Kit
Supplier: PROMEGA SUNNYVALE INC SE
Wizard®
A1120 FF3760
470203-860EA 279.4 USD InStock
470203-860 470203-866
The Wizard® Genomic Plant DNA Purification Kit
Educational Classroom Kits and Activities
A simple method to isolate genomic DNA from plants.
  • Extract plant DNA with a simple, 60-minute scalable protocol
The Wizard® Genomic DNA Purification Kit provides a simple, solution-based method for isolation of DNA. DNA purified with this system is suitable for a variety of applications, including amplification, digestion with restriction endonucleases and membrane hybridizations (e.g., Southern and dot/slot blots).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty