Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Abbott™ Alere™ hCG Combo Cassette (20/10 mIU/ml), Healthcare Products
Supplier: National Distribution and Contracting
Abbott™ Alere™
MPC92215
76553-910EA 90.28 USD InStock
76553-910
Abbott™ Alere™ hCG Combo Cassette (20/10 mIU/ml), Healthcare Products
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Alere™ hCG combo cassette (25 mIU/ml) is a one-step test for the early detection of human chorionic gonadotropin (hCG), a hormone produced during pregnancy.
  • Fast - Single-step test provides positive results in as little as 3 minutes for urine and 5 minutes for serum
  • Easy - CLIA-Waived (for urine), easy-to-use test
  • Reliable - Highly sensitive assay with built-in control ensures accuracy
  • Flexible - Test can be used with either urine or serum samples
The test uses a patented, innovative technology to provide highly sensitive and specific results for pregnancy. A sensitivity of 25 mIU/ml in urine and serum offers clear, consistent results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty