Would you like to log in again?
Abbott™ BinaxNOW™ G6PD Test Kit, Healthcare Products
Supplier: Healthcare Products
BinaxNOW®
MPC780-000
76553-970EA 1003.95 USD InStock
76553-970
Abbott™ BinaxNOW™ G6PD Test Kit, Healthcare Products
Assays Immunological Assays
The BinaxNOW™ G6PD test is a rapid test for the qualitative detection of G6PD enzyme activity in human venous whole blood. The simple, three step test provides results in less than 10 minutes, allowing for prompt treatment decisions.
The BinaxNOW™ G6PD (Glucose-6-Phosphate Dehydrogenase) Test is an in vitro enzyme chromatographic test for the qualitative detection of G6PD enzyme activity in human venous whole blood, collected in heparin or ethylenediaminetetraacetic acid (EDTA).
The BinaxNOW™ G6PD Test is a visual screening test used for differentiating normal from deficient G6PD activity levels in whole blood and is intended to aid in the identification of people with G6PD deficiency. Samples which generate deficient results should be assayed using a quantitative G6PD test method to verify the deficiency.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty