Would you like to log in again?
Abbott™ Alere™ hCG Dipstick, Healthcare Products
Supplier: National Distribution and Contracting
Abbott™ Alere™
MPC92211
76553-926EA 99.84 USD InStock
76553-926
Abbott™ Alere™ hCG Dipstick, Healthcare Products
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Alere™ hCG dipstick test provides qualitative determination of human chorionic gonadotropin (hCG) in urine for early detection of pregnancy.
  • Fast - Single-step test provides positive results in as little as 3 minutes
  • Easy - CLIA-waived, easy-to-use test
A simple procedure provides easy-to-read results in 3 minutes at a detection level of 25 mIU/ml.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty