Would you like to log in again?
CiDecon® II ST Concentrated Phenolic Liquid Disinfectant, Sterile, Decon Labs
Supplier: Decon Labs
8512
76461-870CS 459.62 USD InStock
76461-870
CiDecon® II ST Concentrated Phenolic Liquid Disinfectant, Sterile, Decon Labs
Cleaning Agents Disinfectants
CiDecon® II ST sterile, phenolic-based liquid disinfecant is packaged in a single serve 2 oz. bottle.
  • Sterile, phenolic-based concentrated disinfecant
  • Effective against a wide range of bacteria, viruses, and fungi
  • Product is irradiated and tested to confirm sterilty
CiDecon® II ST is a double-bagged, gamma irradiated, concentrated, phosphate-free, detergent-enhanced disinfectant formulated for use in cleanroom areas and surfaces where broad-based disinfection is required. The multi-phenolic formula is designed to clean, disinfect, and deodorize any washable, inanimate, non porous surface in one easy step. Dilution is one ounce per gallon, and this single serve two ounce bottle is used with two gallons of water.
CiDecon® II ST is gamma-irradiated to a sterility assurance level of 10⁻⁶. Each lot is tested for sterility via a USP 14-day sterility test prior to release. A lot specific document detailing QC, irradiation, and sterility is shipped with each case. CiDecon® II ST can be alternated with LopHene II ST if a rotational disinfection program is desired, or where organism resistance is suspected.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty