Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Eisco Burette Tip for Eisco Pinch Clip Burettes
Supplier: EISCO SCIENTIFIC LLC
CH0234TIP
470344-528EA 3.49 USD InStock
470344-528
Eisco Burette Tip for Eisco Pinch Clip Burettes
Burets
Replacement tip for Eisco pinch clip burettes.
Single burette tip made of high quality 3.3 borosilicate glass. For use with Eisco pinch clip burettes only. Tip measures 2.7" long, has a body diameter of 7 mm and a tip diameter of 3 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty