Would you like to log in again?
Panthera TEC Material Trinocular Microscope (BF/RL 6×4 Stage) Packages, Motic
Supplier: Motic
MBPANMT2-S6 MBPANMT2-1080X MBPANMT2-4000
76614-984EA 6605 USD InStock
76614-984 76614-982 76614-986
Panthera TEC Material Trinocular Microscope (BF/RL 6×4 Stage) Packages, Motic
Microscopes
The Panthera TEC offers high value for the inspection of semiconductors and various other materials, especially in industrial quality control and technical education.
  • Panthera TEC-BF/RL 6×4 stage (trino)
  • 0.5× C-Mount camera adapter for ¹/₃" and ¹/₂" chip sensors
  • Motic analyzes basic 64-bit; USB + dongle (no charge)
  • Moticam S6/1080×/BMH4000
Brightfield, darkfield and cross-polarization contrast are combined with a new, unique-to-Motic segmental illumination that all contribute to the versatility of this model. This concept allows an oblique incident illumination, perfect for the detection of scratches or other defects on flat and reflecting surfaces without a need to move the sample. BD models feature LD Plan BD objectives with a superb imaging performance, mounted on a fivefold nosepiece.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty