Would you like to log in again?
PA43 BIO Trinocular Routine Lab Microscope Packages (Standard), Motic
Supplier: Motic
PA43BIOT-1080X PA43BIOT-S6 PA43BIOT-4000
76615-048EA 7621 USD InStock
76615-048 76615-050 76615-052
PA43 BIO Trinocular Routine Lab Microscope Packages (Standard), Motic
Microscopes
The new PA43 BIOMED is a new series from Motic Scientific and represents a large step forward in combining modularity with ergonomics to improve application imaging workflows. A progression into routine work, this new microscope’s robust design and solid footprint does not compromise on optical quality and features.
  • PA43BIO trino upright compound routine lab microscope
  • 0.5× C-mount camera adapter for ¹/₃" and ¹/₂" chip sensors
  • Motic analyzes basic 64-bit; USB + dongle (no charge)
  • Moticam S6/1080X/BMH4000
Its modularity offers flexibility when choosing a configuration that suits multiple end-user applications from cost-effective solutions to high-quality optical systems for lab diagnostics. The microscope is not only modular but also flexible in design and can be upgraded at any time.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty