Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 40% (w/w)

Supplier: Ricca Chemical
Danger
Synonyms: Potassium hydrateCaustic potash

R6180000-20F
RCR6180000-20FEA 1277.2 USD InStock
RCR6180000-20F
Potassium hydroxide 40% (w/w)
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Boiling Pt: ~100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 39.95 - 40.05 % (w/w)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty