Would you like to log in again?
FlexTech™ Y9299 Cut-Resistant Sandy Nitrile Palm Coated Gloves, Wells Lamont
Supplier: Wells Lamont
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
FlexTech™
Y9299XXL Y9299L Y9299S Y9299M Y9299XL
76595-902DZ 111.15 USD InStock
76595-902 76595-890 76595-906 76595-888 76595-900
FlexTech™ Y9299 Cut-Resistant Sandy Nitrile Palm Coated Gloves, Wells Lamont
Gloves
Protect hands with ANSI A5 cut resistance and a sandy nitrile palm.
  • 18 gauge thin glove for better dexterity
  • Touchscreen compatible
The Y9299 glove series offers ANSI Cut level A5 protection to reduce hand injuries, while providing increased abrasion resistance for extra durability and prolonged use. The ultra-lightweight liner provides a comfortable fit and excellent dexterity. The black sandy nitrile palm coating ensures a secure grip in wet, dry, or oily applications.
Certifications: ANSI level A5 cut resistance. ANSI Puncture level 4. ANSI abrasion level 4.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty