Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eco-Heat Heating Mantle

Supplier: United Scientific Supplies
UNEHMTLE-250
470334-202EA 134.25 USD InStock
470334-202
Eco-Heat Heating Mantle
Heaters Heating Mantles
Economical heating mantle for all your laboratory needs.

  • Compact design
  • For round bottom boiling flasks up to 250 ml
  • Heats up to 380 °C

Continuously adjustable heating from ambient room temperature to a maximum of 380 °C. Mantle is woven from glass fiber yarn to provide uniform heating to round bottom flasks. The inner core is constructed of high temperature E-glass fiber that protects against corrosion and provides durability and high temperature resistance.

Technical specifications:
• Heating function: Electronic thermoregulation
• Heating power: 100 - 2400 W
• Maximum heating temperature: 380 °C
• Operates on 115V/60 Hz

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty