Would you like to log in again?
Investigation Rocks and Minerals Chart
2575
470347-362BX 0.6 USD InStock
470347-362
Investigation Rocks and Minerals Chart
Charts
Hand made 3D geologic posters built to investigate and identify rocks and minerals.
  • Rock identification
  • 93 of the most common rocks and minerals
  • For display and instruction
  • Grades 5 and up
93 of the most common mineral and rock specimens are ordered and classified by their composition and how they are formed on the investigating minerals and rock chart.
The chart highlights 40 minerals, 18 igneous rocks, 18 sedimentary rocks, and 17 metamorphic rocks. All are mounted on an 18×24" wood framed chart which is perfect for use in all educational environments! Each specimen is identified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty