Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Tribromobenzene

Supplier: APOLLO SCIENTIFIC
Warning

OR4123-100G OR4123-25G OR4123-500G
76583-272EA 37 USD InStock
76583-272 76583-274 76583-270
1,3,5-Tribromobenzene
1,3,5-Tribromobenzene


Formula: C₆H₃Br₃
MW: 314.8 g/mol
Boiling Pt: 271 °C
Melting Pt: 117…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00000080
CAS Number: 626-39-1
EINECS: 210-947-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty