Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: p-Bromobenzoic Acid

OR4997-500G OR4997-100G OR4997-25G
76586-004EA 69 USD InStock
76586-004 76586-002 76586-000
4-Bromobenzoic acid 98%
4-Bromobenzoic acid


Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 252…258 °C
Density: 1.894 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002529
CAS Number: 586-76-5
EINECS: 209-581-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty