Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-1,3-dimethoxybenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Bromo-2,4-dimethoxybenzene

OR4515-100G OR4515-25G
76584-544EA 56 USD InStock
76584-544 76584-542
4-Bromo-1,3-dimethoxybenzene 98%
4-Bromo-1,3-dimethoxybenzene


Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 153…155 °C/18mmHg
Melting Pt: 24…26 °C
Density: 1.507 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009844
CAS Number: 17715-69-4
EINECS: 241-717-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty