Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR4976-100G OR4976-500G OR4976-25G
76585-970EA 56 USD InStock
76585-970 76585-972 76585-968
2-Nitrobenzaldehyde 98%
2-Nitrobenzaldehyde


Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 153 °C/23mmHg
Melting Pt: 42…46 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty