Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloroanisole

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Chloro-4-methoxybenzene

OR4349-250G OR4349-50G
76583-754EA 36 USD InStock
76583-754 76583-756
4-Chloroanisole
4-Chloroanisole


Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 198…202 °C
Melting Pt: –18 °C
Density: 1.164 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00000616
CAS Number: 623-12-1
EINECS: 210-772-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty