Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitrobenzyl alcohol 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (3-Nitrophenyl)methanol

OR5016-25G OR5016-100G
76586-168EA 47 USD InStock
76586-168 76586-166
3-Nitrobenzyl alcohol 99%
3-Nitrobenzyl alcohol


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 175…180 °C/3mmHg
Melting Pt: 30…32 °C
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007273
CAS Number: 619-25-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty