Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',3'-Dichloroacetophenone 95+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-(2,3-Dichlorophenyl)ethanone

OR8271-5G OR8271-25G OR8271-100G
76588-668EA 29 USD InStock
76588-668 76588-672 76588-670
2',3'-Dichloroacetophenone 95+%
2',3'-Dichloroacetophenone


Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 106…108 °C/2.0mmHg
Density: 1.293 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00052988
CAS Number: 56041-57-7
EINECS: 259-954-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty