Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dichlorobenzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (2,4-Dichlorophenyl)methanol

OR5024-100G OR5024-500G OR5024-25G
76586-200EA 246 USD InStock
76586-200 76586-198 76586-196
2,4-Dichlorobenzyl alcohol 97%
2,4-Dichlorobenzyl alcohol


Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 150 °C/25mmHg
Melting Pt: 54…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004606
CAS Number: 1777-82-8
EINECS: 217-210-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty