Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrobenzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (4-Nitrophenyl)methanol

OR5015-1KG OR5015-500G OR5015-100G
76586-160EA 16 USD InStock
76586-160 76586-164 76586-162
4-Nitrobenzyl alcohol 98%
4-Nitrobenzyl alcohol


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 185 °C/12mmHg
Melting Pt: 92…95 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007376
CAS Number: 619-73-8
EINECS: 210-611-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty