Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Bromoacetanilide 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-BromoacetanilideN-(4-Bromophenyl)acetamide

OR4282-100G
76583-680EA 53 USD InStock
76583-680
4'-Bromoacetanilide 99%
4'-Bromoacetanilide


Formula: H₃CC(O)NH(C₆H₄Br)
MW: 214.06 g/mol
Melting Pt: 165…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00000092
CAS Number: 103-88-8
EINECS: 203-154-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty