Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Iodoacetophenone

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: p-Iodoacetophenone1-(4-Iodophenyl)ethan-1-one1-(4-Iodophenyl)ethanone

OR7749-5G OR7749-100G OR7749-25G
76588-216EA 57 USD InStock
76588-216 76588-214 76588-212
4'-Iodoacetophenone
4'-Iodoacetophenone


Formula: C₈H₇IO
MW: 246.05 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty