Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylurea

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: N-Ethylurea

OR42229-25G OR42229-500G OR42229-100G
76583-484EA 70 USD InStock
76583-484 76583-486 76583-488
Ethylurea
Ethylurea


Formula: C₃H₈N₂O
MW: 88.11 g/mol
Melting Pt: 87…90 °C
Flash Pt: 93.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00007953
CAS Number: 625-52-5
EINECS: 210-898-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty