Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitrobenzoylacetonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR7010-1G OR7010-5G
76587-464EA 13 USD InStock
76587-464 76587-466
3-Nitrobenzoylacetonitrile
3-Nitrobenzoylacetonitrile


Formula: C₉H₆N₂O₃
MW: 190.16 g/mol
Storage Temperature: Ambient
CAS Number: 21667-64-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty