Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Tolylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Dihydroxy(4-methylphenyl)boranep-Tolylboronic acid4-Methylbenzeneboronic acid(4-Methylphenyl)metaboric acid4-Methylphenylboronic acid

OR6888-25G OR6888-100G OR6888-5G
76587-222EA 13.39 USD InStock
76587-222 76587-224 76587-226
4-Tolylboronic acid
4-Tolylboronic acid


Formula: C₇H₉BO₂
MW: 135.96 g/mol
Melting Pt: 256…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00039138
CAS Number: 5720-05-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty