Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Isonipecotic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Isonipecotic acidPiperidine-4-carboxylic acid(±)-4-Piperidinecarboxylic acid4-Piperidinecarboxylic acid

OR6923-1KG OR6923-25G OR6923-100G
76587-254EA 120 USD InStock
76587-254 76587-250 76587-252
(±)-Isonipecotic acid
(±)-Isonipecotic acid


Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006004
CAS Number: 498-94-2
EINECS: 207-872-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty