Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-pyridineboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 6-Bromopyridine-3-boronic acid6-Bromo-3-pyridinylboronic acid2-Bromo-5-pyridinylboronic acid2-Bromopyridine-5-boronic acid(6-Bromopyridin-3-yl)boronic acid

OR7164-1G OR7164-5G OR7164-10G
76587-720EA 13.39 USD InStock
76587-720 76587-722 76587-724
2-Bromo-5-pyridineboronic acid 98%
2-Bromo-5-pyridineboronic acid


Formula: C₅H₅BBrNO₂
MW: 201.81 g/mol
Melting Pt: 161…166 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD03411558
CAS Number: 223463-14-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty