Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobenzaldehyde dimethyl acetal

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Bromo-4-(dimethoxymethyl)benzene4-Bromo-α,α-dimethoxytoluene

OR70059-25G OR70059-100G
76587-440EA 13 USD InStock
76587-440 76587-442
4-Bromobenzaldehyde dimethyl acetal
4-Bromobenzaldehyde dimethyl acetal


Formula: C₉H₁₁BrO₂
MW: 231.09 g/mol
Boiling Pt: 254 °C
Density: 1.383 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD01320499
CAS Number: 24856-58-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty