Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichloro-4-pyridone

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3,5-Dichloropyridin-4(1H)-one

OR7551-5G OR7551-10G OR7551-1G OR7551-25G
76588-042EA 14 USD InStock
76588-042 77424-234 76588-040 76588-044
3,5-Dichloro-4-pyridone
3,5-Dichloro-4-pyridone


Formula: C₅H₃Cl₂NO
MW: 163.9894 g/mol
Melting Pt: 325 °C
Storage Temperature: Ambient
MDL Number: MFCD00051673
EINECS: 241-267-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty