Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Benzyloxy)benzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR52207-100G OR52207-500G
76587-056EA 29 USD InStock
76587-056 76587-058
2-(Benzyloxy)benzaldehyde 98%
2-(Benzyloxy)benzaldehyde


Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 326 °C
Density: 1.339 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016583
CAS Number: 5896-17-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty