Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(2-Tetrahydrofuroyl)piperazine 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (Piperazin-1-yl)(tetrahydrofuran-2-yl)methanone

OR6858-25G OR6858-100G OR6858-10G
76587-176EA 101.63 USD InStock
76587-176 76587-178 76587-174
1-(2-Tetrahydrofuroyl)piperazine 97%
1-(2-Tetrahydrofuroyl)piperazine


Formula: C₉H₁₆N₂O₂
MW: 184.24
Boiling Pt: 120…125 °C/0.2mmHg
Density: 1.17 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00190247
CAS Number: 63074-07-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty