Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-1-Boc-3-hydroxypiperidine 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate(3S)-3-Hydroxypiperidine, N-BOC protected

OR4581-25G OR4581-5G OR4581-100G
76584-768EA 13.39 USD InStock
76584-768 76584-772 76584-770
(S)-1-Boc-3-hydroxypiperidine 98%
(S)-1-Boc-3-hydroxypiperidine


Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 34…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115307
CAS Number: 143900-44-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty