Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-chlorobenzaldehyde 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR46035-25G OR46035-100G OR46035-5G
76584-844EA 371 USD InStock
76584-844 76584-842 76584-840
2-Bromo-6-chlorobenzaldehyde 99%
2-Bromo-6-chlorobenzaldehyde


Formula: C₇H₄BrClO
MW: 219.46 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00266792
CAS Number: 64622-16-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty