Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-6-chloroimidazo[1,2-b]pyridazine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR7413-25G OR7413-10G OR7413-250MG
76587-954EA 13.39 USD InStock
76587-954 76587-956 76587-958
3-Bromo-6-chloroimidazo[1,2-b]pyridazine 98%
3-Bromo-6-chloroimidazo[1,2-b]pyridazine


Formula: C₆H₃BrClN₃
MW: 232.47 g/mol
Melting Pt: 122…126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027274
CAS Number: 13526-66-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty