Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Morpholino)phenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (4-Morpholinophenyl)boronic acid

OR7242-1G OR7242-5G OR7242-250MG
76587-810EA 13.39 USD InStock
76587-810 76587-812 76587-814
4-(Morpholino)phenylboronic acid
4-(Morpholino)phenylboronic acid


Formula: C₁₀H₁₄BNO₃
MW: 207.0341 g/mol
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD03095169
CAS Number: 186498-02-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty