Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-N-Boc-1,4-cyclohexanediamine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: trans-tert-Butyl (4-aminocyclohexyl)carbamatetrans-4-(Boc-amino)cyclohexylamine

OR7177-25G OR7177-5G OR7177-1G
76587-740EA 113 USD InStock
76587-740 76587-736 76587-738
trans-N-Boc-1,4-cyclohexanediamine
trans-N-Boc-1,4-cyclohexanediamine


Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
Melting Pt: 74.5…75.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03001719
CAS Number: 177906-48-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty