Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Benzylhydroxylamine hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR51891-5G OR51891-25G
76586-850EA 16 USD InStock
76586-850 76586-848
N-Benzylhydroxylamine hydrochloride 97%
N-Benzylhydroxylamine hydrochloride


Formula: C₇H₁₀ClNO
MW: 159.6134 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00043377
CAS Number: 29601-98-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty