Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Di-tert-butyl-2,2'-bipyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4,4'-Di-tert-butyl-2,2'-bipyridine4,4'-Di-tert-butyl-2,2'-bipyridyl

OR70014-5G OR70014-10G OR70014-1G
77423-972EA 72 USD InStock
77423-972 76587-358 76587-360
4,4'-Di-tert-butyl-2,2'-bipyridine 98%
4,4'-Di-tert-butyl-2,2'-bipyridine


Formula: C₁₈H₂₄N₂
MW: 268.4000 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863731
CAS Number: 72914-19-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty